ADVH49B824BF AChemBlock

6-chloro-[1,2,4]triazolo[4,3-a]pyridine-3-carboxylic acid

  • Cas Number : 1159831-32-9
  • Molecular Weight : 197.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-1-2-4-triazolo-4-3-a-pyridine-3-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S82992
Availability details for
Quantity
  • Availability
  • Add to Cart