ADVH49B824C0 AChemBlock

methyl 6-bromo-[1,2,4]triazolo[1,5-a]pyridine-2-carboxylate

  • Cas Number : 1159811-32-1
  • Molecular Weight : 256.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-bromo-1-2-4-triazolo-1-5-a-pyridine-2-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83004
Availability details for
Quantity
  • Availability
  • Add to Cart