ADVH49B824C2 AChemBlock

2-fluoro-3-(trifluoromethoxy)benzenamine

  • Cas Number : 1159512-64-7
  • Molecular Weight : 195.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-fluoro-3-trifluoromethoxy-benzenamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71812
Availability details for
Quantity
  • Availability
  • Add to Cart