ADVH49B824C4 AChemBlock

5-{[(2R,3R,4R,5R,6R)-4,5-bis(acetyloxy)-6-[(acetyloxy)methyl]-3-acetamidooxan-2-yl]oxy}pentanoic acid

  • Cas Number : 1159408-54-4
  • Molecular Weight : 447.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-2r-3r-4r-5r-6r-4-5-bis-acetyloxy-6-acetyloxy-methyl-3-acetamidooxan-2-yl-oxy-pentanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83017
Availability details for
Quantity
  • Availability
  • Add to Cart