ADVH49B824C6 AChemBlock

(6-(methoxycarbonyl)-1H-indol-2-yl)boronic acid

  • Cas Number : 1150114-47-8
  • Molecular Weight : 219

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methoxycarbonyl-1h-indol-2-yl-boronic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83165
Availability details for
Quantity
  • Availability
  • Add to Cart