ADVH49B824CB AChemBlock

2,4,6-trichloro-5-iodo-pyrimidine

  • Cas Number : 1137576-61-4
  • Molecular Weight : 309.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-4-6-trichloro-5-iodo-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83207
Availability details for
Quantity
  • Availability
  • Add to Cart