ADVH49B824D7 AChemBlock

4-bromo-1,6-dimethyl-pyridin-2-one

  • Cas Number : 1114563-01-7
  • Molecular Weight : 202.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-1-6-dimethyl-pyridin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83324
Availability details for
Quantity
  • Availability
  • Add to Cart