ADVH49B824E7 AChemBlock

1-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl]piperidin-1-yl}ethan-1-one

  • Cas Number : 1092563-66-0
  • Molecular Weight : 319.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-4-4-tetramethyl-1-3-2-dioxaborolan-2-yl-1h-pyrazol-1-yl-piperidin-1-yl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83447
Availability details for
Quantity
  • Availability
  • Add to Cart