ADVH49B824E9 AChemBlock

methyl 3,5-dichloro-1-methyl-1H-pyrazole-4-carboxylate

  • Cas Number : 109056-69-1
  • Molecular Weight : 209.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-5-dichloro-1-methyl-1h-pyrazole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83476
Availability details for
Quantity
  • Availability
  • Add to Cart