ADVH49B824EA AChemBlock

4-bromo-5-methoxy-benzene-1,2-diamine

  • Cas Number : 108447-01-4
  • Molecular Weight : 217.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-bromo-5-methoxy-benzene-1-2-diamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83499
Availability details for
Quantity
  • Availability
  • Add to Cart