ADVH49B824EC AChemBlock

(2,3-dihydro-1-benzofuran-6-yl)methanol

  • Cas Number : 1083168-69-7
  • Molecular Weight : 150.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-3-dihydro-1-benzofuran-6-yl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83514
Availability details for
Quantity
  • Availability
  • Add to Cart