ADVH49B824EF AChemBlock

Methyl 6-chloro-1H-indole-4-carboxylate

  • Cas Number : 1082040-57-0
  • Molecular Weight : 209.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-chloro-1h-indole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72419
Availability details for
Quantity
  • Availability
  • Add to Cart