ADVH49B824F3 AChemBlock

4-(Pyridin-4-ylmethyl)-2H-phthalazin-1-one

  • Cas Number : 107558-48-5
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-pyridin-4-ylmethyl-2h-phthalazin-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86707
Availability details for
Quantity
  • Availability
  • Add to Cart