ADVH49B824F5 AChemBlock

3-Amino-2-chloropyridine-5-boronic acid, pinacol ester

  • Cas Number : 1073354-96-7
  • Molecular Weight : 272.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link https://www.achemblock.com/3-amino-2-chloropyridine-5-boronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70740
Availability details for
Quantity
  • Availability
  • Add to Cart