ADVH49B824F7 AChemBlock

2,3-Difluorophenylboronic acid pinacol ester

  • Cas Number : 1073339-17-9
  • Molecular Weight : 258.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/2-3-difluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70479
Availability details for
Quantity
  • Availability
  • Add to Cart