ADVH49B824FD AChemBlock

1-tert-butyl 2-methyl 2,3-dihydro-1H-pyrrole-1,2-dicarboxylate

  • Cas Number : 1063989-43-4
  • Molecular Weight : 227.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-tert-butyl-2-methyl-2-3-dihydro-1h-pyrrole-1-2-dicarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83622
Availability details for
Quantity
  • Availability
  • Add to Cart