ADVH49B824FE AChemBlock

4-chloro-5-hydroxy-pyridine-2-carboxylic acid

  • Cas Number : 1060804-56-9
  • Molecular Weight : 173.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-chloro-5-hydroxy-pyridine-2-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83648
Availability details for
Quantity
  • Availability
  • Add to Cart