ADVH49B82502 AChemBlock

Methyl 4-amino-3-bromo-5-nitrobenzenecarboxylate

  • Cas Number : 105655-17-2
  • Molecular Weight : 275.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-amino-3-bromo-5-nitrobenzenecarboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86492
Availability details for
Quantity
  • Availability
  • Add to Cart