ADVH49B82507 AChemBlock

4-Carboxy-2-fluorophenylboronic acid pinacol ester

  • Cas Number : 1050423-87-4
  • Molecular Weight : 284.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-carboxy-2-fluorophenylboronic-acid-pinacol-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S70432
Availability details for
Quantity
  • Availability
  • Add to Cart