ADVH49B82509 AChemBlock

tert-butyl (5-bromo-1,3,4-thiadiazol-2-yl)carbamate

  • Cas Number : 1048358-33-3
  • Molecular Weight : 280.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-5-bromo-1-3-4-thiadiazol-2-yl-carbamate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86503
Availability details for
Quantity
  • Availability
  • Add to Cart