ADVH49B8250C AChemBlock

IRAK inhibitor 6

  • Cas Number : 1042672-97-8
  • Molecular Weight : 396.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/irak-inhibitor-6.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86744
Availability details for
Quantity
  • Availability
  • Add to Cart