ADVH49B8250D AChemBlock

(S)-6-fluoro-1-isopropyl-3,4-dihydronaphthalen-2(1H)-one

  • Cas Number : 104205-01-8
  • Molecular Weight : 206.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-6-fluoro-1-isopropyl-3-4-dihydronaphthalen-2-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86482
Availability details for
Quantity
  • Availability
  • Add to Cart