ADVH49B8250E AChemBlock

methyl 6-amino-1H-indole-4-carboxylate

  • Cas Number : 103956-00-9
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-6-amino-1h-indole-4-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83792
Availability details for
Quantity
  • Availability
  • Add to Cart