ADVH49B82516 AChemBlock

6-bromoisoquinolin-3-ol

  • Cas Number : 1031927-91-9
  • Molecular Weight : 224.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromoisoquinolin-3-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71853
Availability details for
Quantity
  • Availability
  • Add to Cart