ADVH49B82519 AChemBlock

5-methylimidazo[1,2-a]pyrimidine-7-carboxylic acid

  • Cas Number : 1030017-70-9
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methylimidazo-1-2-a-pyrimidine-7-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83885
Availability details for
Quantity
  • Availability
  • Add to Cart