ADVH49B8251E AChemBlock

4-methoxybenzene-1,2-diamine

  • Cas Number : 102-51-2
  • Molecular Weight : 138.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methoxybenzene-1-2-diamine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71859
Availability details for
Quantity
  • Availability
  • Add to Cart