ADVH49B8251F AChemBlock

3-bromo-1,5,6,7-tetrahydro-1,8-naphthyridine

  • Cas Number : 1023813-80-0
  • Molecular Weight : 213.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-1-5-6-7-tetrahydro-1-8-naphthyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S83922
Availability details for
Quantity
  • Availability
  • Add to Cart