ADVH49B82523 AChemBlock

3-(Dimethylamino)-2-(1H-1,2,3-triazol-1-yl)acrylic acid methyl ester

  • Cas Number : 1021869-28-2
  • Molecular Weight : 196.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/3-dimethylamino-2-1h-1-2-3-triazol-1-yl-acrylic-acid-methyl-ester.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86703
Availability details for
Quantity
  • Availability
  • Add to Cart