ADVH49B82526 AChemBlock

6-bromo-[1,2,4]triazolo[4,3-a]pyrimidine

  • Cas Number : 1019023-12-1
  • Molecular Weight : 199.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-bromo-1-2-4-triazolo-4-3-a-pyrimidine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86504
Availability details for
Quantity
  • Availability
  • Add to Cart