ADVH49B82528 AChemBlock

Potassium 1-methyl-4-trifluoroboratomethylpiperazine

  • Cas Number : 1015484-22-6
  • Molecular Weight : 220.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/potassium-1-methyl-4-trifluoroboratomethylpiperazine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72352
Availability details for
Quantity
  • Availability
  • Add to Cart