ADVH49B8252C AChemBlock

3-phenyl-1,3-thiazolidine-2,4-dione

  • Cas Number : 1010-53-3
  • Molecular Weight : 193.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/3-phenyl-1-3-thiazolidine-2-4-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S86603
Availability details for
Quantity
  • Availability
  • Add to Cart