ADVH49B8252D AChemBlock

methyl (3R,6S)-6-methylpiperidine-3-carboxylate

  • Cas Number : 1009376-81-1
  • Molecular Weight : 157.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3r-6s-6-methylpiperidine-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84139
Availability details for
Quantity
  • Availability
  • Add to Cart