ADVH49B8252F AChemBlock

6-chloro-N-isopropylpyridazin-3-amine

  • Cas Number : 1007-55-2
  • Molecular Weight : 171.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/6-chloro-n-isopropylpyridazin-3-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71872
Availability details for
Quantity
  • Availability
  • Add to Cart