ADVH49B82530 AChemBlock

1-methoxycyclopropane-1-carboxylic acid

  • Cas Number : 100683-08-7
  • Molecular Weight : 116.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-methoxycyclopropane-1-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84165
Availability details for
Quantity
  • Availability
  • Add to Cart