ADVH49B82533 AChemBlock

2-(6-methylpyridin-3-yl)ethan-1-ol

  • Cas Number : 100189-17-1
  • Molecular Weight : 137.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-6-methylpyridin-3-yl-ethan-1-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84252
Availability details for
Quantity
  • Availability
  • Add to Cart