ADVH49B82537 AChemBlock

5-Methoxy-6-methyl-1H-indazole

  • Cas Number : 1000343-61-2
  • Molecular Weight : 162.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-methoxy-6-methyl-1h-indazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72398
Availability details for
Quantity
  • Availability
  • Add to Cart