ADVH49B82538 AChemBlock

3-Iodo-4-methoxy-5-azaindole

  • Cas Number : 1000341-25-2
  • Molecular Weight : 274.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-iodo-4-methoxy-5-azaindole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72412
Availability details for
Quantity
  • Availability
  • Add to Cart