ADVH49B82539 AChemBlock

3-Bromo-4-methoxy-5-azaindole

  • Cas Number : 1000341-22-9
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-methoxy-5-azaindole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72432
Availability details for
Quantity
  • Availability
  • Add to Cart