ADVH49B8253A AChemBlock

2-amino-5-bromopyridine-4-carboxylic acid

  • Cas Number : 1000339-23-0
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-amino-5-bromopyridine-4-carboxylic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S71880
Availability details for
Quantity
  • Availability
  • Add to Cart