ADVH49B82540 AChemBlock

4-nitrocyclopentene

  • Molecular Weight : 113.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/4-nitrocyclopentene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84306
Availability details for
Quantity
  • Availability
  • Add to Cart