ADVH49B82547 AChemBlock

7-bromo-2,3,4,5-tetrahydro-1,4-benzoxazepin-5-one

  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/7-bromo-2-3-4-5-tetrahydro-1-4-benzoxazepin-5-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84313
Availability details for
Quantity
  • Availability
  • Add to Cart