ADVH49B82548 AChemBlock

3-(2-hydroxyethyl)cyclohexanone

  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-2-hydroxyethyl-cyclohexanone.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84315
Availability details for
Quantity
  • Availability
  • Add to Cart