ADVH49B8254C AChemBlock

5-hydroxy-1,2-dihydropyridin-2-one

  • Molecular Weight : 111.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-hydroxy-1-2-dihydropyridin-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84320
Availability details for
Quantity
  • Availability
  • Add to Cart