ADVH49B8254D AChemBlock

1H-imidazol-4-amine

  • Molecular Weight : 83.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1h-imidazol-4-amine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84321
Availability details for
Quantity
  • Availability
  • Add to Cart