ADVH49B82550 AChemBlock

2-amino-3-cyclobutylpropanoic acid

  • Molecular Weight : 143.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-amino-3-cyclobutylpropanoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84324
Availability details for
Quantity
  • Availability
  • Add to Cart