ADVH49B8255C AChemBlock

bicyclo[4.2.0]octa-1,3,5-trien-7-one

  • Molecular Weight : 118.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/bicyclo-4-2-0-octa-1-3-5-trien-7-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84335
Availability details for
Quantity
  • Availability
  • Add to Cart