ADVH49B8255D AChemBlock

5-amino-6-methylpyridine-3-carbonitrile

  • Molecular Weight : 133.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/5-amino-6-methylpyridine-3-carbonitrile.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84336
Availability details for
Quantity
  • Availability
  • Add to Cart