ADVH49B8255F AChemBlock

6-methoxy-1,2,3,4-tetrahydronaphthalene-2-carbaldehyde

  • Molecular Weight : 190.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/6-methoxy-1-2-3-4-tetrahydronaphthalene-2-carbaldehyde.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84338
Availability details for
Quantity
  • Availability
  • Add to Cart