ADVH49B82561 AChemBlock

1H,4H,5H,6H-cyclopenta[c]pyrazole

  • Molecular Weight : 108.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1h-4h-5h-6h-cyclopenta-c-pyrazole.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S84340
Availability details for
Quantity
  • Availability
  • Add to Cart