ADVH49B82564 AChemBlock

TRANS-tetrahydrofuran-3,4-dicarboxylic acid

  • Molecular Weight : 160.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/trans-tetrahydrofuran-3-4-dicarboxylic-acid-295753.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S72443
Availability details for
Quantity
  • Availability
  • Add to Cart